Rolling cabinets for ROTINA 420 / 420 R centrifuges, Hettich Lab Technology
Fournisseur: HETTICH INSTRUMENTS
Certificats
À propos de cet article
For convenient storage and positioning. Cabinets for centrifuges save space on the laboratory bench by providing enhanced flexibility.
- Save valuable bench space
- Store accessories in one location
- Ergonomic height for loading/unloading
The cabinets lower the centrifuge to an ergonomic height for easy loading and unloading. Each cabinet
provides ample storage for user manuals, rotors, buckets and accessories. Important documentation and accessories remain with the centrifuge at all times. All cabinets are of sturdy design and feature casters with two wheel locks, drawer locks and two keys.
Frequently Bought Together