Electrode Storage Bottle, Lamotte | Avantor
Electrode Storage Bottle, Lamotte
Electrode Soaker Bottle, 20 ml
Catalog # 66170-856
Supplier:   LaMotte
Fournisseur #:  0668
Electrode Storage Bottle, Lamotte
Catalog # 66170-856
Supplier:  LaMotte
CAS Number:  
Electrode Storage Bottle, Lamotte
Electrode Soaker Bottle, 20 ml
CAS Number:  
Price
$48.63
Log in pour la disponibilité
Each
Product Details & Documents
Continuously soaks pH electrode in a storage solution to prevent probe dry out.
Twist top O ring seal prevents leaks.
Spécifications
Spécifications
  • Désignation:
    Electrode Soaker Bottle
  • Volume:
    20 ml