Evaporating Dishes, with Lip, CoorsTek
Catalog # 25313-072
Supplier: Coors Ceramics
Specifications
- Capacity:90 ml
- Cat. no.:25313-072
- Diameter ×H:90×22 mm
- Supplier No.:60232
Specifications
About this item
Evaporating with lip, shallow form, glazed inside except for rim and partially outside.