Flying Pig
Flying Pig
Catalog # 470223-114
Supplier: ARBOR SCIENTIFIC
Fournisseur #: P4-2165
Jump to:
Product Details & Documents
Make students learn when pigs fly.
- Includes mounting string
- Includes instruction manual
Add some excitement to your lessons on circular motion with the Flying Pig. Suspended from the ceiling, his wings flap and carry him in a regular circle. With the string, this motion gives students a conical pendulum and a perfect opportunity for quantitative measurements of circular motion.
Ordering information: Lab instructions included.
Spécifications
- Désignation:Flying Pig