24/Seven™ Edging for Modular Floor Mats, Wearwell®
Supplier: Wearwell
Certificates
About this item
24/Seven is a collection of high performance, 3x3’, interconnecting rubber anti-fatigue mats.
- Ability to replace single tiles saves money and down time
- Hidden interlocks
- Available in 3 different rubber compounds
Interlocking rubber tiles easily configure for custom work station ergonomics. Choose from three compounds: GR natural rubber for dry areas and exposure to water, an innovative CFR compound resistant to chemicals and mineral oil based cutting fluids, or NBR for areas with petrochemicals.
Accessories information: 24/Seven has optional 3" wide safety edges. Male and Female ramps are available in all three compounds.
Frequently Bought Together