Midi Plus™ Pipetting Controller, Sartorius
Midi Plus™ Pipetting Controller, Sartorius
Supplier:   Sartorius
Midi Plus™ Pipetting Controller, Sartorius
Supplier:  Sartorius
Certificates
Midi Plus™ Pipetting Controller, Sartorius
Midi Plus™ Pipetting Controller, Sartorius
1 Options Available Below
Product Details & Documents
The lightweight, cordless Midi Plus pipetting controller enables users to aspirate from bottles and tubes without holding their arms and hands in an uncomfortable raised position.
  • Precise finger tip control of variable speed ranges
  • Hydrophobic autoclavable filter prevents over aspiration
  • Low-battery warning
Users can achieve fine-tune speed by applying varying finger pressure to the operating buttons.
The pipetting controller fits all commonly used 1 to 100 ml glass or plastic pipettes.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Midi Plus™ Pipetting Controller
    Discontinued
    Supplier Number: 710931
  • Midi Plus™ Pipetting Controller
    Log in for availability
    Log in for availability
    Each
    Each - $974.99
    $974.99
    Each
       $974.99
    Minimum order quantity is 1
    Supplier Number: 710931
Tips
Product Information
  • Catalog #
    Longueur
    Tip Style
    Sterile
    Volume
    Availability
    Price
  • 242.0 mm
    Standard
    -
    500 - 10000 µl
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $131.69
    $131.69
    Case of 100
       $131.69
Product Information
  • Catalog #
    Longueur
    Tip Style
    Sterile
    Volume
    Availability
    Price
  • 150.0 mm
    Standard
    -
    100 - 5000 µl
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $62.40
    $62.40
    Case of 100
       $62.40
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Adapter Set for Midi Plus™, Standard F/1 to 100 mL
    Adapter Set for Midi Plus™, Standard F/1 to 100 mL
    75840-134
    Log in for availability
    Each - $294.87
Product Information
  • Catalog #
    Availability
    Price
  • Adapter Set for Midi Plus™, Plus F/10 mL
    Adapter Set for Midi Plus™, Plus F/10 mL
    75840-138
    Log in for availability
    Each - $298.73