Deluxe Tuf Sponge Anti-Fatigue Mat, Wearwell®
: Wearwell
This blown-vinyl sponge mat is ergonomically designed to reduce floor hardness by up to 50%.
- Economic comfort for light industrial applications
- Safety rounded edges
- Standard lengths or easily cut to fit on-site
Deluxe Tuf Sponge Mats are a fantastic choice for practicality and economy. This blown-vinyl sponge is ergonomically designed to reduce floor hardness by up to 50%. Rolls of Tuf Sponge can easily be custom cut to fit any work area. This thicker and softer formulation feels great and is specifically designed to reduce standing worker fatigue. Ideally suited for use in dry environments.
: Meets ASTM F1677. Dry: COF = 0.94/Wet: COF = 0.72. Meets ASTM D575/0.276 @ 35 psi.
Frequently Bought Together