VAI® ALCOH-WIPE® Wipes, Veltek Associates
Wipes saturated with 70% USP isopropyl alcohol and 30% USP water for injection.
- Validated for sterility and shelf life
- 100% synthetic filament constructed in a continuous knit that minimizes fiber and particulate release
Use to wipe down filling and packing machinery and for spill control on lexan, glass, polycarbonate, and stainless steel.
: Available as individual packs, 100 per case.
Frequently Bought Together