VWR® Rectangular Carboy, Wide Mouth
À propos de cet article
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
- Non sterile
- Polypropylene and HDPE formats available
- Clear graduation marks
- Excellent chemical resistance
- Wide mouth
- Heavy duty stainless steel handle for easy carrying and pouring
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. HDPE carboys are not autoclavable.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
3 Options Available Below
Product Details & Documents
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
- Non sterile
- Polypropylene and HDPE formats available
- Clear graduation marks
- Excellent chemical resistance
- Wide mouth
- Heavy duty stainless steel handle for easy carrying and pouring
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. HDPE carboys are not autoclavable.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Product Family Options
Informations sur le produit
- DésignationCapacitéMatièreNeck I.D.AvailabilityPrice
- Carboy, Wide Mouth, Rectangular, Autoclavable10 LPP100 mmLog in pour la disponibilitéEach(1/Itm/Bx)Each(1/Itm/Bx) - $208.94$208.94Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 6 (1/ Itm/Bx)Case of 6 (1/ Itm/Bx) - $1,253.62$1,253.62Prix promotionnel pour les nouveaux clientsEach(1/Itm/Bx)$208.94Case of 6 (1/ Itm/Bx)$1,253.62
- Carboy, Wide Mouth, Rectangular, Autoclavable20 LPP100 mmLog in pour la disponibilitéEach(1/Itm/Bx)Each(1/Itm/Bx) - $300.98$300.98Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 4 (1/ Itm/Bx)Case of 4 (1/ Itm/Bx) - $1,203.94$1,203.94Prix promotionnel pour les nouveaux clientsEach(1/Itm/Bx)$300.98Case of 4 (1/ Itm/Bx)$1,203.94
- Carboy, Wide Mouth, Rectangular20 LHDPE100 mmLog in pour la disponibilitéEach(1/Itm/Bx)Each(1/Itm/Bx) - $525.44$525.44Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 4 (1/ Itm/Bx)Case of 4 (1/ Itm/Bx) - $2,101.76$2,101.76Prix promotionnel pour les nouveaux clientsEach(1/Itm/Bx)$525.44Case of 4 (1/ Itm/Bx)$2,101.76