Accutech® 91-225 Sterile Rubber Latex Gloves, Powder-Free, Ansell
Fournisseur: Microflex
Certificats
À propos de cet article
Comfortable cleanroom gloves with low allergenic content and smooth grip.
- Inner silicone coating facilitates easy donning and double gloving
- Thinner fingertips and bisque finish provide greater sensitivity, increased handling precision, and dexterity
- Low allergenic content minimizes the risk of latex-associated allergies for the wearer
Certifications: EN 374, Cat. III, CE marked.
Packaging: One pair per poly wallet, sealed in a poly envelope; 10 pairs per sealed polybag; 5 sealed polybags per master polybag; 4 master polybags per lined carton; 200 pairs per carton.
Spécifications
- Catalog No:
- 10818-414
- 10818-412
- 10818-418
- 10818-416
- 10818-406
- 10818-410
- 10818-408
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight