HardyDisks™ AST Cefaclor, CEC-30, Hardy Diagnostics
Catalog # 75846-316
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Cefaclor, CEC-30, HDx
- Size:1x50 disks
- Cat. no.:75846-316
- Supplier No.:Z8121
Specifications
About this item
Cefaclor, CEC-30, HardyDisks™ AST, for susceptibility testing.
- Impregnated paper disks used for Antimicrobial Susceptibility Testing (AST)
- Spring-loaded for smooth and reliable dispensing
- Compatible with single or multi-disk dispensers
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.The last disk in each cartridge is clearly marked so as to alert you when refills are needed.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers as listed below.
Delivery: Freeze on arrival.