ContactUsLinkComponent
LKV Agar, Reducible, Hardy Diagnostics
Catalog # 75846-312
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
LKV Agar, Reducible, Hardy Diagnostics
Catalog # 75846-312
Supplier:  Hardy Diagnostics
Supplier Number:  A60
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    LKV Agar, HDx
  • Size:
    15x100 mm plate
  • Cat. no.:
    75846-312
  • Supplier No.:
    A60
Specifications
About this item
LKV (Laked Sheep Blood, Kanamycin, Vancomycin) Agar, reducible, for anaerobes.
  • Selective isolation of gram negative anaerobic bacteria
  • Laked Sheep Blood, Kanamycin and Vancomycin agar
  • Reducible, for anaerobes
(Brucella Agar with Laked Sheep Blood, Kanamycin, Vancomycin, Hemin and Vitamin K) For the selective isolation of gram negative anaerobic bacteria, especially Bacteroides spp.