ALA Disks™ Delta-Aminolevulinic Acid, Hardy Diagnostics
Catalog # 75846-306
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:ALA Disks™, Delta-Aminolevulinic Acid
- Size:1x50 disks
- Cat. no.:75846-306
- Supplier No.:Z7081
Specifications
About this item
ALA Disks™, delta-aminolevulinic acid, for the differentiation and speciation of Haemophilus spp., a porphyrin test.
- A useful aid for differentiating species of Haemophilus
- Spring-loaded for smooth and reliable dispensing
- Single metal disk dispenser
Each cartridge contains 50 disks that are spring-loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences.
ALA Disks require the use of a handheld 366 nm ultra violet lamp.