Difco™ Nitrate Broth, BD Life Sciences | Avantor
Difco™ Nitrate Broth, BD Life Sciences
Nitrate Broth, 500 g
Catalog # CA90001-916
Supplier:   BD
Fournisseur #:  226810
Difco™ Nitrate Broth, BD Life Sciences
Catalog # CA90001-916
Supplier:  BD
CAS Number:  
Difco™ Nitrate Broth, BD Life Sciences
Difco™ Nitrate Broth, BD Life Sciences
Nitrate Broth, 500 g
CAS Number:  
Price
619,00 $
Log in pour la disponibilité
Each (500g)
Product Details & Documents
Nitrate Broth is recommended as an aid in the identification of aerobic and facultative anaerobic gram-negative microorganisms by means of the nitrate reduction test.
  • Basal medium containing potassium nitrate
  • Addition of zinc dust is used to detect unreduced nitrate
  • Organisms derive oxygen from nitrate through anaerobic respiration
The formation of nitrogen gas, an end product typical of certain organisms, is evidenced by displacement of the medium from the Durham tube by the gas produced.
Spécifications
Spécifications
  • Désignation:
    Nitrate Broth
  • Taille:
    500 g