Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Difco™ Malt Extract Broth, BD Diagnostics

Difco™ Malt Extract Broth, BD Diagnostics

Malt Extract Broth, 500 g
Catalog # CA90000-150
Supplier:   BD
Supplier #:  211320
Difco™ Malt Extract Broth, BD Diagnostics
Catalog # CA90000-150
Supplier:  BD
CAS Number:  
Difco™ Malt Extract Broth, BD Diagnostics
Difco™ Malt Extract Broth, BD Diagnostics

Malt Extract Broth, 500 g

CAS Number:  
Price
$265.00
Log in for availability
Each (500g)
Product Details & Documents

Malt Extract Broth is used for cultivating yeasts and molds.

  • Malt extract provides carbon, protein, and nutrient sources for essential growth
  • Maltose is an energy source
  • Dextrose is included as a source of fermentable carbohydrate
  • Yeast extract provides vitamins and additional nitrogen and carbon

The acidic pH of Malt Extract Broth allows for the optimal growth of molds and yeasts while restricting bacterial growth.

Specifications

Specifications

  • Désignation:
    Malt Extract Broth
  • Taille:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.