BBL™ Nutrient Broth, BD Life Sciences
Nutrient Broth, 5 mL
  CA90001-450
 :   BD
 :  221669
BBL™ Nutrient Broth, BD Life Sciences
  CA90001-450
 :  BD
 :  
BBL™ Nutrient Broth, BD Life Sciences
Nutrient Broth, 5 mL
 :  
$48.50
Pack of 10 (5ml)
Nutrient Broth is a general-purpose medium for the cultivation of nonfastidious bacteria.
  • Peptone and beef extract provide essential nutrients
  • Very simple nutrient formula
  • Effective for nonfastidious microorganism growth
This relatively simple formulation supports the growth of nonfastidious microorganisms due to its content of peptone and beef extract.
  • Désignation:
    Nutrient Broth
  • Taille:
    5 mL