ContactUsLinkComponent
Introduction to Oxidation/Reduction (Redox) Student Activity
Introduction to Oxidation/Reduction (Redox) Student Activity
Catalog # 470007-038
Supplier:  Lab-Aids
undefined
Introduction to Oxidation/Reduction (Redox) Student Activity
Catalog # 470007-038
Supplier:  Lab-Aids
Supplier Number:  88
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Introduction to Oxidation/Reduction (Redox) Student Activity
  • No. of Lab Groups:
    12
  • Cat. no.:
    470007-038
  • Includes:
    Teacher's Guide, Student Guides and Worksheets, SDS
  • Supplier No.:
    88
Specifications
About this item
An Excellent Introduction to a Very Important Chemical Concept
  • Explore the exchange of electrons redox reactions
  • Highly visible and safe activity
  • Requires one 50 minutes classroom period to complete
Students place pieces of zinc, lead, and copper in solutions of metallic ions to test their relative reducing ability. Then, they compare and determine the relative oxidizing strength of certain halogen elements: chlorine, bromine, and iodine.
Ordering information: Materials include LAB-AIDS® Chemplates®, Copper Shot, Lead Shot, Zinc Shot, Droppers, Copper II Nitrate Solution, Lead II Nitrate Solution, Zinc Nitrate Solution, Chlorine Water, Bromine Water, Iodine Solution, Organic Solvent, Sodium Bromide Solution, Sodium Chloride Solution, Sodium Iodide Solution.
Delivery: Limited shelf life.