Radioactive Waste Disposal Bags, Research Products
Fournisseur: RPI
Certificats
À propos de cet article
Radioactive material waste bags help sort and identify different types of waste.
Strong, bottom-sealed waste liners are free of heavy metals, making them safe for landfill or incineration.
Frequently Bought Together