ContactUsLinkComponent
Urine Sample Tray, Dynamic Diagnostics
Catalogue # 101614-640
Fournisseur:  DYNAMIC DIAGNOSTICS MS
undefined
Urine Sample Tray, Dynamic Diagnostics
Catalogue # 101614-640
Fournisseur:  DYNAMIC DIAGNOSTICS MS
Supplier Number:  2000-109
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Urine Sample Tray with Label
  • Dimensions:
    18.9 x 13 x 3.3"
  • Cat. no.:
    101614-640
  • Supplier No.:
    2000-109
Spécifications
À propos de cet article
Urine sample tray with label holds up to 24 containers
Outer shell can be separated and used for storage or transportation of laboratory containers.