ContactUsLinkComponent
Nanoject III™ Programmable Nanoliter Injector, Drummond
Catalogue # 490019-810
Fournisseur:  Drummond
undefined
Nanoject III™ Programmable Nanoliter Injector, Drummond
Catalogue # 490019-810
Fournisseur:  Drummond
Supplier Number:  3-000-207
Spécifications
  • Description:
    Nanoject III Programmable Nanoliter Injector (100/240V)
  • Cat. no.:
    490019-810
  • Supplier No.:
    3-000-207
Spécifications
À propos de cet article
The Programmable Nanoject III Nanoliter Injector offers a number of features for easy setup and operation as well as more accurate injection volumes down to 0.6 nanoliters.
  • User-Friendly Programmable Touchscreen Operation
  • Positive Micropipet Attachments
  • No O-Rings Required
  • Easy Setup
  • Hydraulic Technology for Accurate and Consistent Injection Volumes
The programmable touchscreen is user friendly, and the positive micropipet attachments do not require O-rings.
The injector's hydraulic technology provides accurate and consistent injection volumes.
Delivery: The kit contains control box, power source, injector head, injector head cable, 3.5" glass capillaries, 7" glass capillaries, backfilling needle, universal adapter and 4 interchangeable adapter plugs.