Avantor SterilEnz®-II/AT: Pre-Gasketed Sanitary Fittings for Single-Use Systems, Avantor Fluid Handling
Supplier: AVANTOR FLUID HANDLING, LLC
Certificates
About this item
SterilEnz®-II/AT connectors come with a platinum cured, medical grade silicone gasket mechanically attached to the fitting face.
- Standard sanitary clamp fitting, per ASME BPE Specifications
- Class VI Compliant materials; certificate included
- Won't leak or blow-off like stepped barbs can
- Mono-Barb design
- Zero mold parting lines on all critical sealing surfaces
- Superior tooling
- Gamma-stable, 25 - 40 kGY
- For use in irradiated disposable systems
- Autoclaveable at 123 °C for 30 minutes
- Certified free from any animal derived components or processes
- Animal-free
SterilEnz®-II/AT fittings are made rugged inert polypropylene suitable for either autoclaving or gamma-irradiation. The new II/AT series is available in seven fitting sizes.
Certifications: USP <88>: Class VI Biological reactivity tests, in vivo
USP <87>: Cytotoxicity, MEM Elution, in vitro
USP <661>: Physicochemical Test for plastics
USP <1207>: Sterile Product Packaging, hose barb integrity evaluation, via microbial immersion
ASTM F756-00 : Rabbit blood cell hemolysis
SterilEnz®-II/AT are certified ADCF (animal derived component free).
All SterilEnz®-II/AT products are assembled inside a certified ISO 7 (Class 10000) environment.
Protected by U.S. Patent 7,950,700, other patents pending. SterilEnz® is a registered trademark of PAW BioScience Products, LLC.
USP <87>: Cytotoxicity, MEM Elution, in vitro
USP <661>: Physicochemical Test for plastics
USP <1207>: Sterile Product Packaging, hose barb integrity evaluation, via microbial immersion
ASTM F756-00 : Rabbit blood cell hemolysis
SterilEnz®-II/AT are certified ADCF (animal derived component free).
All SterilEnz®-II/AT products are assembled inside a certified ISO 7 (Class 10000) environment.
Protected by U.S. Patent 7,950,700, other patents pending. SterilEnz® is a registered trademark of PAW BioScience Products, LLC.
Specifications
- Catalog No:
- 75838-672
- 75838-670
- 75838-674
- 75838-678
- 75838-676
- 75838-680
- 75838-668
- Max. operating pressure:
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- Max. temperature:
- –20...+75 °C
- –20...+75 °C
- –20...+75 °C
- –20...+75 °C
- –20...+75 °C
- –20...+75 °C
- –20...+75 °C