Qualatex® XC 8C Static Dissipative Latex Fingercots, QRP
: PIP
The QRP 8C Static Dissipative Latex Finger Cots have an average surface resistance of 10⁷ ohms per square surface unit.
- Packaged for ISO 5 (Class 100) applications
- Compliant with ASTM and IEST-RP-CC 005.3 standards
- Average surface resistance of 10⁷ ohms per square surface unit
- Fore use with Class I and Class II static sensitive devices
- Unique, Stand-Up pouch - easy product identification
The QRP 8C Static Dissipative Latex Finger Cots have an average surface resistance of 10⁷ ohms per square surface unit. Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers. Compliant with ASTM and IEST-RP-CC 005.3 standards. For use with Class II static sensitive devices (thresholds above 1000V). Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2. Unique, Stand-Up Pouch provides easy product identification.
: ESD Safe to ANSO/ESD S20.20-2014.
Frequently Bought Together