Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers.
- Packaged for ISO 5 (Class 100) applications.
- Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers
- Unique, Stand-Up Pouch- easy product identification
The 5C Qualatex® powder-free cleanroom latex finger cots are packaged for ISO 5 (Class 100) applications
Applications: Discrete components, hard drive assemblies, die bonding, wire attach, disk drives/thin film heads, qoptics, finishing, cleaning & handling of precision instruments.
Compliant with ASTM and IEST-RP-CC 005.3 standards. Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers. Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2. Unique, Stand-Up Pouch provides easy product identification.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.