Qualatherm® 59G Dry Thermal Protection Gloves, 14", QRP
Fournisseur: PIP
Certificats
À propos de cet article
QRP 59G Qualatherm gloves are RoHS compliant gloves, packaged for ISO 5 Class 100 cleanroom applications.
- Dry contact temperatures from –210 to 1000 °F (–134 to 537 °C) with no charring, ash, or residue
- No PCB, asbestos, or fiberglass
Applications: Diffusion furnaces, quartz wafer boards, liquid gas flask handling (ultra cold), hot/dry glassware handling and metal fabrication.
The middle layer is 100% virgin wool knit with a high terry pile for excellent insulation. Completely nylon-lined to minimize particulate shed during donning and removal.
Spécifications
- Catalog No:
- 89041-062
- 32889-019
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5