Qualatex® 2C Low Powder Latex Finger Cots, QRP
Fournisseur: PIP
Certificats
À propos de cet article
QRP 2C Finger Cots provide natural latex protection from skin salts, flakes, and oils, dyes, chemical, and solvent splash.
- For general , personal, and product protection
- Sheer film for maximum dexterity
- Economical
- For industrial use only
High level of comfort, abrasion resistance, and tactile sensitivity. Textured, rolled, nominal 2.75" length. 3 mil, tissue weight, sheer film for maximum dexterity and relief from finger fatigue.
Caution: This product contains natural rubber latex which may cause allergic reactions.
Frequently Bought Together