Orion™ Automatic Stirrer Probe and Paddle, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Use the Orion™ Star Stirrer Probe with Orion™ benchtop meters to stir solutions for better accuracy without the difficulty of using a stir plate and bar.
Use the Orion™ Star Stirrer Probe with Orion™ Versa Star™ meters, Orion™ Star A210 series benchtop meters, and legacy Orion™ Star series benchtop meters to stir standards and samples for better measurement accuracy without the difficulty of separate stir plate and bars. The Orion™ Star Stirrer Probe is powered and controlled by the meter with five optimized stirring speeds. The device fits in the electrode stand for easy movement in and out of samples and can be cleaned simply by rinsing. Paddles can be replaced to extend the life of Orion™ Star Stirrer Probes.
Frequently Bought Together