To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Lysing Matrix I, 2 mL Tubes
- Size:100×2 ml tubes
- Format:Ceramic Spheres, Single tubes; no cross contamination
- Adapter compatibility:QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- Instrument Compatibility:FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
- Tube compatibility:2 ml
- Key applications:Extractions of nucleic acids, proteins, metabolites and other small molecules from tough, hard or brittle cell membranes including organisms with a tough or dense exterior matrix. Isolation of intact organelles. Breaking exoskeleton of arthropods and crustaceans. Dry Grinding fungal spores
- Material of construction:Matrix I, contains 2 mm yellow zirconia beads and a 4 mm black ceramic beads
- Molecule enriched:DNA; RNA; Proteins; Other small molecules
- Materials required (not supplied):FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- Storage and handling:Store at Room Temperature (15...30 °C)
- Sample volume:up to 200 mg per prep
- Sample type:Plants; Animal Tissues; Wood; Seeds; Soil; Clay; Ancient and Dried Samples; Spores
- Hardness:Yellow Zirconium Oxide Beads, Vickers 900 - 1000; 4 mm ceramic bead, Mohs 7.5
- Specific gravity:Yellow Zirconium Oxide Beads, 5.5; 4 mm ceramic bead, 6.0 - 6.25
- Product type:Lab Essentials
- Cat. no.:CA76046-188
Specifications
About this item
Each impact-resistant 2 mL tube contains 2 mm yellow zirconia beads and a 4 mm black ceramic sphere.
- High intertia for lysis by impaction
- High impaction but low shearing
- Lyse tough chitin/protein composites
Zirconium oxide has very high density and high tensile strength and hardness, and is effective at breaking very tough and hard samples, including organisms within a tough or dense exterior matrix. This is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing. The 4 mm ceramic sphere provides very high inertia for lysis by impaction of tough chitin/protein composites that comprise the exoskeleton of arthropods and crustaceans. Matrix I can also be used for dry grinding fungal spores to isolate RNA and Proteins.