Order Entry
Canada
ContactUsLinkComponent
CoolTeenPrep™ Adapter, MP Biomedicals, LLC
CoolTeenPrep™ Adapter, MP Biomedicals, LLC
  CA76046-180
undefined
CoolTeenPrep™ Adapter, MP Biomedicals, LLC
  CA76046-180
 :  116002530
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Capacity:
    6 Tubes
  • Description:
    CoolTeenPrep™ 6×15 ml Adapter
  • Size:
    6×15 ml
  • Sample throughput:
    Low, Medium
  • Adapter compatibility:
    FastPrep®-24 Adapters, Cryogenic, Large Volume
  • Instrument Compatibility:
    FastPrep-24; FastPrep-96 (requires ConeFlex™ Legacy Adapter)
  • Kit compatibility:
    All miniprep 15 ml
  • Tube compatibility:
    15 ml
  • Key applications:
    Extractions of nucleic acids, proteins, metabolites and other small molecules from virtually any temperature unstable or sensitive sample. Larger sized Sample Holder
  • Material of construction:
    Composite
  • Storage and handling:
    Store at Room Temperature (15...30 °C)
  • Sample volume:
    up to 12 g
  • Sample type:
    Soil; Plants; Animal Tissues; Feces; Yeast; Bacteria (Gram +/-); Fungi; Spores; Wastewater; Food; Seeds; Roots
  • Weight:
    627 g
  • Product type:
    Instruments & Systems
  • Cat. no.:
    CA76046-180

 

 

CoolTeenPrep™ 6×15 ml Adapter is ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.

  • Consistent, reproducible results
  • For use with FastPrep-24 or FastPrep-96 Homogenizer
  • Ensure efficient cooling of samples

CoolTeenPrep™ 6×15 ml Adapter is a cryogenic adapter which allows simultaneous lysis of 6×15 ml samples. Based on passive temperature control technology, the CoolTeenPrep™ adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray, is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep® precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. A wide range of disposable 15 ml Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.