To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:24 Tubes
- Description:CoolPrep™ 24×2 ml Adapter
- Size:24×2 ml
- Sample throughput:Low, Medium
- Adapter compatibility:FastPrep®-24 Adapters, Cryogenic, High-Throughput
- Instrument Compatibility:FastPrep-24; FastPrep-96 (requires ConeFlex™ Legacy Adapter)
- Kit compatibility:FastDNA and FastDNA Spin; FastRNAPro; FastRNA Spin; FastProtein; GeneClean for Ancient DNA; all miniprep 2 ml
- Tube compatibility:2 ml
- Key applications:Extractions of nucleic acids, proteins, metabolites and other small molecules from virtually any temperature unstable or sensitive sample
- Material of construction:Composite
- Storage and handling:Store at Room Temperature (15...30 °C)
- Sample volume:up to 5 g
- Sample type:Soil; Plants; Animal Tissues; Feces; Yeast; Bacteria (Gram +/-); Fungi; Spores; Wastewater; Food; Seeds; Roots
- Weight:680 g
- Product type:Instruments & Systems
- Cat. no.:CA76046-178
Specifications
About this item
CoolPrep™ Adapter is ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.
- Consistent, reproducible results
- For use with FastPrep-24 or FastPrep-96 Homogenizer
- Ensure efficient cooling of samples
CoolPrep™ 24×2 ml Adapter is a cryogenic adapter which allows simultaneous lysis of 24×2 ml samples. Based on passive temperature control technology, the CoolPrep™ adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep® precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. Most samples can be processed in 40 seconds or less.