MetaPhor® Agarose | Avantor
MetaPhor® Agarose
Supplier:   Lonza
MetaPhor® Agarose
Supplier:  Lonza
Certificates
MetaPhor® Agarose
MetaPhor® Agarose
3 Options Available Below
Product Details & Documents
MetaPhor® is an intermediate melting temperature agarose that lets you resolve DNA fragments differing in size by 2%, in the range of 200 bp to 800 bp.
  • All Lonza agarose is quality tested to certify performance
  • High gel strength
  • Standard melting temperature
MetaPhor® Agarose is a high resolution agarose that challenges polyacrylamide. MetaPhor® Agarose is an intermediate melting temperature (75 °C) agarose with twice the resolution capabilities of the finest-sieving agarose products. For example, a 200 bp DNA fragment can be separated from a 204 bp fragment. Using fast running protocols DNA differing in size by 1% can be resolved in as little as 1.5 hours in a 20 cm long horizontal or vertical gel format. MetaPhor agarose gels (2 to 4%) approximate the resolution of polyacrylamide gels (4 to 8%). These agarose gels are ideal for resolving AMPFLPs, STRs, and tri- and tetranucleotide repeats. Ideal for small PCR fragments, MetaPhore® is the choice agarose for resolving hard to see bands.
Applications: Small fragments, PCR.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Utilisation
    Température de stockage
    Availability
    Price
  • MetaPhor® agarose
    25 g
    2% DNA resolution between 200 and 800 bp; ≤600 kDa
    18...26 °C (64...79 °F)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 360,05 $
    360,05 $
    Each
       360,05 $
    La quantité minimale de commande est de 1
    Supplier Number: 50181
  • MetaPhor® agarose
    125 g
    2% DNA resolution between 200 and 800 bp; ≤600 kDa
    18...26 °C (64...79 °F)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (125g)
    Each (125g) - 1 234,52 $
    1 234,52 $
    Each (125g)
       1 234,52 $
    La quantité minimale de commande est de 1
    Supplier Number: 50180
  • MetaPhor® agarose
    500 g
    2% DNA resolution between 200 and 800 bp; ≤600 kDa
    18...26 °C (64...79 °F)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500g)
    Each (500g) - 3 942,85 $
    3 942,85 $
    Each (500g)
       3 942,85 $
    La quantité minimale de commande est de 1
    Supplier Number: 50184