Costar® Transtar-96® Liquid Transfer System, Corning
96 Well Liquid Transfer System, 25 - 200 µl, ±5%
Catalog # 29442-530
Supplier: Corning
Supplier #: 7605
Jump to:
Product Details & Documents
The Costar® Transtar-96® System is a portable, autoclavable liquid handling system designed for use with 96 well plates.
- Ideal for changing cell culture media and screening monoclonal antibodies
- Enables liquids to be aspirated, transferred and dispensed
Specifications
- Volume:25 - 200 µl
- Désignation:96 Well Liquid Transfer System
- Accuracy:±5%
Accessories
Product Information
- Catalog #AvailabilityPrice
- Costar® Transtar-96® Disposable Reservoir Liner, Open, SterileCostar® Transtar-96® Disposable Reservoir Liner, Open, Sterile29442-524Log in for availabilityCase of 100 - 272,16 $
Product Information
- Catalog #AvailabilityPrice
- Costar® Transtar-96® Disposable Reservoir Liner, 12 channel, SterileCostar® Transtar-96® Disposable Reservoir Liner, 12 channel, Sterile29442-526Log in for availabilityCase of 100 - 266,94 $
Product Information
- Catalog #AvailabilityPrice
- Costar® Transtar-96® Disposable Reservoir Liner, 8 channel, SterileCostar® Transtar-96® Disposable Reservoir Liner, 8 channel, Sterile29442-528Log in for availabilityCase of 100 - 311,00 $