VWR® Basic Industrial Nylon Bouffants
À propos de cet article
VWR® Basic nylon bouffant caps provide lightweight, breathable head coverage for controlled environments. Made from durable nylon, these caps are designed to minimize contamination by keeping hair securely contained. The non-linting material helps maintain cleanroom integrity, while the elastic closure ensures a comfortable, secure fit for various head sizes.
Durable nylon for long-lasting performance
- Lightweight and breathable material ensures comfort during extended wear
- Non-linting design maintains cleanroom and controlled environment integrity
- Elastic closure provides a secure and comfortable fit for various head sizes
- Latex-free construction is safe for users with latex sensitivities
- Manufactured in ISO-certified facilities for consistent quality
VWR® Basic Nylon Bouffant Caps are designed to provide reliable head coverage in cleanrooms, laboratories, and other controlled environments where contamination control is essential. Made from durable nylon, these caps offer a lightweight and breathable solution that ensures comfort during extended wear. The non-linting material helps maintain the integrity of sensitive environments by minimizing particle generation, making them ideal for applications requiring strict cleanliness standards.
Each cap features an elastic closure that provides a secure and comfortable fit for various head sizes, ensuring convenience and ease of use. Latex-free construction makes these bouffant caps suitable for individuals with latex sensitivities, while their disposable design offers a cost-effective alternative to reusable headwear, reducing laundering requirements and contamination risks. Manufactured in ISO certified facilities and lot controlled for consistent quality, VWR® Basic Nylon Bouffant Caps deliver dependable performance for contamination sensitive operations.
Packaging: Bouffant caps are packaged 1000 per case.
9 Options Available Below
Product Details & Documents
VWR® Basic nylon bouffant caps provide lightweight, breathable head coverage for controlled environments. Made from durable nylon, these caps are designed to minimize contamination by keeping hair securely contained. The non-linting material helps maintain cleanroom integrity, while the elastic closure ensures a comfortable, secure fit for various head sizes.
Durable nylon for long-lasting performance
- Lightweight and breathable material ensures comfort during extended wear
- Non-linting design maintains cleanroom and controlled environment integrity
- Elastic closure provides a secure and comfortable fit for various head sizes
- Latex-free construction is safe for users with latex sensitivities
- Manufactured in ISO-certified facilities for consistent quality
VWR® Basic Nylon Bouffant Caps are designed to provide reliable head coverage in cleanrooms, laboratories, and other controlled environments where contamination control is essential. Made from durable nylon, these caps offer a lightweight and breathable solution that ensures comfort during extended wear. The non-linting material helps maintain the integrity of sensitive environments by minimizing particle generation, making them ideal for applications requiring strict cleanliness standards.
Each cap features an elastic closure that provides a secure and comfortable fit for various head sizes, ensuring convenience and ease of use. Latex-free construction makes these bouffant caps suitable for individuals with latex sensitivities, while their disposable design offers a cost-effective alternative to reusable headwear, reducing laundering requirements and contamination risks. Manufactured in ISO certified facilities and lot controlled for consistent quality, VWR® Basic Nylon Bouffant Caps deliver dependable performance for contamination sensitive operations.
Packaging: Bouffant caps are packaged 1000 per case.
Documents
Certificates
Recherche par numéro de lotSpecifications for Product Family
Spécifications
- Catalog No:
- 75997-646
- 75829-204
- 75997-636
- 76076-778
- 76076-776
- 76076-780
- 76076-784
- 76076-782
- 76076-786
- Fabric:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Food|Industrial|Laboratory|Cleanroom
- Food|Industrial|Laboratory|Cleanroom
- Food|Industrial|Laboratory|Cleanroom
- Food|Industrial|Laboratory|Cleanroom
- Food|Industrial|Laboratory|Cleanroom
- Food|Industrial|Laboratory|Cleanroom
- Food|Industrial|Laboratory|Cleanroom
- Food|Industrial|Laboratory|Cleanroom
- Food|Industrial|Laboratory|Cleanroom
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Water resistance:
- First line barrier
- First line barrier
- First line barrier
- First line barrier
- First line barrier
- First line barrier
- First line barrier
- First line barrier
- First line barrier
- Weight:
- 10 g/m²
- 10 g/m²
- 10 g/m²
- 10 g/m²
- 10 g/m²
- 10 g/m²
- 10 g/m²
- 10 g/m²
- 10 g/m²
Spécifications
Product Family Options
18" bouffant caps
Informations sur le produit
- ColorØAvailabilityPrice
- White45.7 cm (18")Log in pour la disponibilitéLog in pour la disponibilitéCase of 1,000Case of 1,000 - $158.57$158.57Prix promotionnel pour les nouveaux clientsCase of 1,000$158.57Spécifications:
More
Specifications
Cat. No.75997-646FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m² - Blue45.7 cm (18")Log in pour la disponibilitéCase of 1,000Case of 1,000 - $152.62$152.62Prix promotionnel pour les nouveaux clientsCase of 1,000$152.62Each$152.62Spécifications:
More
Specifications
Cat. No.76076-778FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m² - Black45.7 cm (18")Case of 1,000$42.97Each$101.58Spécifications:
More
Specifications
Cat. No.76076-784FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m²
21" bouffant caps
Informations sur le produit
- ColorØAvailabilityPrice
- White53.3 cm (21")Log in pour la disponibilitéLog in pour la disponibilitéCase of 1,000Case of 1,000 - $93.07$93.07Prix promotionnel pour les nouveaux clientsCase of 1,000$93.07Spécifications:
More
Specifications
Cat. No.75829-204FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m² - Blue53.3 cm (21")Log in pour la disponibilitéCase of 1,000Case of 1,000 - $163.22$163.22Prix promotionnel pour les nouveaux clientsCase of 1,000$163.22Each$163.22Spécifications:
More
Specifications
Cat. No.76076-776FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m² - Black53.3 cm (21")Log in pour la disponibilitéCase of 1,000Case of 1,000 - $91.37$91.37Prix promotionnel pour les nouveaux clientsCase of 1,000$91.37Each$91.37Spécifications:
More
Specifications
Cat. No.76076-782FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m²
24" bouffant caps
Informations sur le produit
- ColorØAvailabilityPrice
- White61 cm (24")Log in pour la disponibilitéLog in pour la disponibilitéCase of 1,000Case of 1,000 - $179.71$179.71Prix promotionnel pour les nouveaux clientsCase of 1,000$179.71Spécifications:
More
Specifications
Cat. No.75997-636FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m² - Blue61 cm (24")Log in pour la disponibilitéCase of 1,000Case of 1,000 - $173.46$173.46Prix promotionnel pour les nouveaux clientsCase of 1,000$173.46Each$173.46Spécifications:
More
Specifications
Cat. No.76076-780FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m² - Black61 cm (24")Log in pour la disponibilitéCase of 1,000Case of 1,000 - $97.29$97.29Prix promotionnel pour les nouveaux clientsCase of 1,000$97.29Each$97.29Spécifications:
More
Specifications
Cat. No.76076-786FabricNylonCleanroom class10000/ISO 7GenderUnisexGarment applicationFood|Industrial|Laboratory|CleanroomParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight10 g/m²