ContactUsLinkComponent
Amine Column Test Mix (GC) (8 Components), Restek
Amine Column Test Mix (GC) (8 Components), Restek
Catalog # 75835-326
Supplier:  Restek
CAS Number:  
Amine Column Test Mix (GC) (8 Components), Restek
Catalog # 75835-326
Supplier:  Restek
Supplier Number:  35002
CAS Number:  
Specifications
  • Concentration:
    In methylene chloride:methanol (1:1)
  • Description:
    Amine Column Test Mix
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75835-326
  • Supplier No.:
    35002
Specifications
About this item
The amine column test mix consists of (C10) decane (0,60 mg/ml), (C12) dodecane (0.60 mg/ml, 1,2-butanediol (0.60 mg/ml), diethanolamine (DEA) (1.20 mg/ml), diethylenetriamine (1.20 mg/ml), 2,6-dimethylaniline (0.60 mg/ml), 2-nonanol (0.60 mg/ml), pyridine(0.60 mg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.