QuEChERS Performance Standard C, Restek
QuEChERS Performance Standard C, 300 µg/ml each in acetonitrile:acetic acid (99.9:0.1), 1 ml/Ampule
Catalog # 75834-630
Supplier:   Restek
 :  31155
QuEChERS Performance Standard C, Restek
Catalog # 75834-630
Supplier:  Restek
CAS Number:  
QuEChERS Performance Standard C, Restek
QuEChERS Performance Standard C, 300 µg/ml each in acetonitrile:acetic acid (99.9:0.1), 1 ml/Ampule
CAS Number:  
Price
$197.04
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Designed for use in all QuEChERS methods for pesticides in fruits and vegetables, including the original unbuffered method, AOAC 2007.01, and EN15662
  • Ideal for initial method evaluations and ongoing method performance validations.
  • Optimized blend of chemically compatible analytes for maximum stability and shelf life.
  • Precise formulation improves data quality and operational efficiency; spend more time running samples and less time sourcing and preparing standards
  • Quantitatively analyzed to confirm the composition and stability of each mixture
  • Produced and tested in accordance with ISO Guide 34 and 17025 accreditation.
 : AOAC 2007.01 and EN15662. Produced and tested in accordance with ISO 17034 and 17025 accreditation.
Spécifications
Spécifications
  • Concentration:
    300 µg/ml each in acetonitrile:acetic acid (99.9:0.1)
  • Désignation:
    QuEChERS Performance Standard C
  • Taille:
    1 ml/Ampule