SP Wilmad-LabGlass Round Bottom Two Vertical Neck Flasks, SP Industries
Fournisseur: SP Industries
Certificats
À propos de cet article
With two vertical necks and a round bottom, these borosilicate glass flasks feature a side neck Standard Taper 24/40 outer joint
The center Standard Taper joint varies with flask capacity.
Frequently Bought Together