EPA 539 Calibration Stock Standard (7 Components), Restek
Catalog # 75834-918
Supplier: Restek
CAS Number:
Specifications
- Matrix:Acetonitrile
- Description:EPA 539 Calibration Stock Standard
- Size:1 ml/Ampule
- Cat. no.:75834-918
- Supplier No.:31998
Specifications
About this item
Contains the seven hormones listed in EPA Method 539 at the appropriate concentrations. The seven components are 4-androstene-3, 17-dione, equilin, 17-beta-Estradiol, estriol, estrone, 17-alpha-ethynylestradiol and testosterone.
Certifications: Contains the seven hormones listed in EPA Method 539 at the appropriate concentrations. The seven components are 4-androstene-3, 17-dione, equilin, 17-beta-Estradiol, estriol, estrone, 17-alpha-ethynylestradiol and testosterone.
Ordering information: UCMR3 method 539 calibration standard with concentrations of 10 to 400 µg/ml to more conveniently match UCMR3 requirements (cat.# 32461) also available.