Student Thermometer | Avantor
Student Thermometer
Student Thermometer
Certificates
Student Thermometer
Student Thermometer
2 Options Available Below
Product Details & Documents
No-roll triangle shape.
  • Partial immersion
  • White backing
  • Features re-usable plastic storage tube
This student grade thermometer is filled with kerosene with red dye.
Product Family Options
Informations sur le produit
  • Range
    Longueur
    Immersion
    Availability
    Price
  • –20...110 C°
    15.2 cm (6")
    Total
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5.05
    $5.05
    Each
       $5.05
    Supplier Number: THPC06
  • −20…+110 °C (0...230 °F)
    30 cm (11¾")
    Partial
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12.80
    $12.80
    Each
       $12.80
    Supplier Number: THPCF1