BBL™ Saline, Normal, BD
Supplier:   BD
BBL™ Saline, Normal, BD
Supplier:  BD
Certificates
BBL™ Saline, Normal, BD
2 Options Available Below
Product Details & Documents
Normal saline is used in procedures that require the use of an isotonic diluent.
  • Ready to use
  • Provide ensured cell integrity
  • Used with a vast array of procedures
An isotonic diluent may be used for dilution of bacterial cells to provide a concentration suitable for microscopic observation, determination of cell numbers, analysis for genetic or metabolic properties, washing cells preparatory to study, or preparation of standardized inocula.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Tube Size
    Availability
    Price
  • BBL™ saline, normal
    100×1 ml
    Size K
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100 (1ml)
    Pack of 100 (1ml) - $300.00
    $300.00
    Pack of 100 (1ml)
       $300.00
    Supplier Number: 297815
  • BBL™ saline, normal
    100×5 ml
    Size K
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100 (5ml)
    Pack of 100 (5ml) - $313.00
    $313.00
    Pack of 100 (5ml)
       $313.00
    Supplier Number: 221819