ContactUsLinkComponent
SP Wilmad-LabGlass Hubbard Specific Gravity Bottle, SP Industries
Catalog # 14202-630
Supplier:  SP Industries
undefined
SP Wilmad-LabGlass Hubbard Specific Gravity Bottle, SP Industries
Catalog # 14202-630
Supplier:  SP Industries
Supplier Number:  LG-3541-100
Specifications
  • Capacity:
    24 ml
  • Dimensions:
    27 Dia. x 72 H mm
  • Cat. no.:
    14202-630
  • Supplier No.:
    LG-3541-100
Specifications
About this item
This borosilicate glass specific gravity bottle is used in standard test methods for specific gravity of a variety of semi-solid materials and emulsions, such as asphalt, cements, road tars, road oils, and soft tar pitches
The solid Standard Taper stopper has a hole approximately 1.6mm in diameter through the center to allow excess sample to escape.
Certifications: Reference: ASTM D 70, AASHO Method T 43.