BBL™ Standard Methods Agar, BD Life Sciences
BBL™ Standard Methods Agar, BD Life Sciences
Supplier:   BD
BBL™ Standard Methods Agar, BD Life Sciences
Supplier:  BD
Certificates
BBL™ Standard Methods Agar, BD Life Sciences
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Standard Methods Agar is used for obtaining microbial plate counts from milk and dairy products, foods, water, and other materials of sanitary importance.
  • Enzymatic digest of casein provides nitrogen and amino acids
  • Yeast extract supplies B-complex vitamins
  • Dextrose is an energy source
  • Agar is the solidifying agent
Standard Methods Agar is made according to the American Public Health Association (APHA) formulation.
Documents
Product Family Options
Product Information
  • Désignation
    Taille
    Availability
    Price
  • BBL™ Standard Methods Agar, Deeps
    20 mL
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $57.60
    $57.60
    Pack of 10
       $57.60
    Supplier Number: 221030
  • BBL™ Standard Methods Agar
    10 x 100 mm; 15 x 100 mm
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $111.00
    $111.00
    Pack of 20
       $111.00
    Supplier Number: 297030
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.