Fluid A, BD Life Sciences | Avantor
Fluid A, BD Life Sciences
Fluid A (Peptone Water, 0.1%), 500 mL
Catalog # CA90005-180
Supplier:   BD
Fournisseur #:  299111
Fluid A, BD Life Sciences
Catalog # CA90005-180
Supplier:  BD
CAS Number:  
Fluid A, BD Life Sciences
Fluid A (Peptone Water, 0.1%), 500 mL
CAS Number:  
Price
$426.00
Log in pour la disponibilité
Pack of 10 (500ml)
Product Details & Documents
Fluid A (peptone water) is used for diluting or rinsing when performing sterility testing.
  • Prepared and ready to use
  • Dilutes or rinses during sterility testing
  • Peptic digest of animal tissue provides source of nitrogen
These fluids aid in the complete rinsing of the membrane filter apparatus and are not toxic to microorganisms.
Documents
Spécifications
Spécifications
  • Désignation:
    Fluid A (Peptone Water, 0.1%)
  • Taille:
    500 mL
Spécifications