Wards® QA Spectroscope | Avantor
Wards® QA Spectroscope
Ward's® QA spectroscope
Catalog # 470016-874
Supplier:   Wards
Fournisseur #:  PH100QAW
Wards® QA Spectroscope
Catalog # 470016-874
Supplier:  Wards
Wards® QA Spectroscope
Wards® QA Spectroscope
Ward's® QA spectroscope
Price
$20.55
Log in pour la disponibilité
Each Case of 10
Product Details & Documents
This semi-quantitative spectroscope is a simple and economical alternative to expensive equipment.
  • Perfect for Chemistry and Physics classrooms
  • Inexpensive and simple to use
  • Instructions included
The Ward's® QA spectroscope has been designed to produce a bright spectrum of visible light decomposed by a diffraction grating, with a clear, easy-to-view translucent scale. Students can analyze spectra from flames and gas emission tubes, measuring wavelengths from 4000 to 7000 Å with an accuracy of +/-50 Å. The spectroscope houses a 37 mm scale, and includes instructions with suggestions for use.
Spécifications
  • Désignation:
    Ward's® QA spectroscope