C-Clamp, Iron
Catalog # 470121-790
Supplier: GSC International
Specifications
- Description:C-Clamp, 1.5" Depth
- Cat. no.:470121-790
- Supplier No.:7-G103
Specifications
About this item
Used for securing support stands to lab benches and for a multitude of other applications.
- Cast metal painted with corrosion resistant paint
- Steel plated screw
- Corrosion resistant
This 3-inch clamp from our collection is built to last, with a sturdy cast metal frame and a steel plated screw that can handle serious clamping power.
The corrosion-resistant paint means you don't have to worry about it rusting, even in damp environments. Whether you're securing a support stand, holding materials in place, or getting creative with your latest STEAM project, this c-clamp is up for the task.