Centurion™ 452 Series Impact Safety Goggles, 3M
: 3M
Centurion™ Safety Impact Goggle is a reliable choice in protective eyewear. Stable, sturdy construction will help protect wearer's eyes.
- Wide-angle wraparound lens and frame for good peripheral vision
- Direct venting system to help minimize fogging
- Accommodates most prescription eyewear
Includes wide nasal flare for added comfort and a direct venting system to minimize fogging. All goggles feature a strap attachment system.
: Meets ANSI Z87.1-2010, CSA Z94.3-2007.
Frequently Bought Together