VWR® Sampling Scoops, PS
VWR® Sampling Scoops, PS
VWR® Sampling Scoops, PS
VWR® Sampling Scoops, PS
Scoops are ideal for the sampling of powder, granules and pastes.
  • Individually packed, gamma radiation sterilized
Special handle construction and the slightly angled edge stands horizontal and can be laid down without the material trickling out.
 : Production, assembling and packaging according to cleanroom Class 7 (10,000). Complies with food and FDA guidelines.
Product Family Options
  • Désignation
    Capacité
    Longueur
    Color
  • Sampling Scoops
    50 ml (1.6 oz.)
    160 mm (6.2")
    White
    Each
    Each - $538.60
    $538.60
    Pack of 100
    Pack of 100 - $427.67
    $427.67
    Each
       $538.60
    Pack of 100
       $427.67
  • Sampling Scoops
    150 ml (5 oz.)
    216 mm (8.5")
    White
    Each
    Each - $779.20
    $779.20
    Pack of 100
    Pack of 100 - $618.78
    $618.78
    Each
       $779.20
    Pack of 100
       $618.78