VWR® Sampling Spoons, PS
Supplier: VWR International
Jump to:
À propos de cet article
Disposable spoons are ideal for the sampling of powder, granules and pastes.
- Individually packed, gamma radiation sterilized
Certifications: Production, assembling and packaging according to cleanroom Class 7 (10,000). Complies with food and FDA guidelines.
2 Options Available Below
Product Details & Documents
Disposable spoons are ideal for the sampling of powder, granules and pastes.
- Individually packed, gamma radiation sterilized
Certifications: Production, assembling and packaging according to cleanroom Class 7 (10,000). Complies with food and FDA guidelines.
Documents
Certificates
Recherche par numéro de lotProduct Family Options
Informations sur le produit
- DésignationLongueurColorCapacitéAvailabilityPrice
- Sampling Spoons12.7 cm (5")White2.5 ml (0.08 oz.)Each$157.55Pack of 100$114.17
- Sampling Spoons17 cm (6.7")White10 ml (0.3 oz.)Log in pour la disponibilitéPack of 100Pack of 100 - $106.40$106.40Prix promotionnel pour les nouveaux clientsEach$134.05Pack of 100$106.40