Large Microscope Slides, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
Certificates
About this item
Large microscope slides are manufactured from 3/4 white glass
These oversized slides feature 90° corners, and measure 5 x 22.8 cm (2 x 9").
Ordering information: Large slides are packaged in a cardboard box, and are individually seperated by white paper.
Frequently Bought Together