Uncharged Microscope Slides, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
À propos de cet article
Microscope slides are manufactured from 3/4 white glass
These uncharged slides are standard size, and feature frosting optimized for hot foil tape labelers such as the Slidemate and Primera.
Ordering information: Slides are packed 72 per cardboard box. Each box is individually sealed. Each case contains 20 boxes.
Frequently Bought Together