CELLROLL Roller Bottle System, Argos Technologies | Avantor
CELLROLL Roller Bottle System, Argos Technologies
Supplier:   Antyila Scientific
CELLROLL Roller Bottle System, Argos Technologies
Supplier:  Antyila Scientific
Certificates
CELLROLL Roller Bottle System, Argos Technologies
CELLROLL Roller Bottle System, Argos Technologies
7 Options Available Below
Product Details & Documents
CELLROLL a reliable cell culture system.
  • Programmable control unit
  • Rotation speeds of either 1-2 rpm or 2-6 rpm
  • Equipped with an alarm system
The modular design of the CELLROLL allows the user to easily scale-up production depending on the needs: one motor unit can control between one and eight bottle racks, each rack accommodating two standard roller bottles. The moisture-resistant motor unit is available with rotation speeds of either 1-2 rpm or 2-6 rpm and is steered by a programmable control unit located outside of the incubator. In case of power failure or accidental disconnection of control and motor unit an alarm system is activated. The corrosion-free and sturdy construction makes the CELLROLL a reliable cell culture system that will keep your cells always "on the move.
Certifications: CE certified.
Ordering information: Includes drive unit, two decks (for 4 roller bottles), a control unit, a power supply and a control cable.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • CELLROLL Complete, 0.1-2.0 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33,422.03
    $33,422.03
    Each
       $33,422.03
    Supplier Number: 13050-00
  • CELLROLL Complete, 2.0-6.0 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32,619.14
    $32,619.14
    Each
       $32,619.14
    Supplier Number: 13050-02
  • CELLROLL Roller System Control Unit
    Discontinued
    Supplier Number: -
  • CELLROLL Drive Unit, 0.1-2.0 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11,239.68
    $11,239.68
    Each
       $11,239.68
    Supplier Number: 24406-27
  • CELLROLL Drive Unit, 2.0-6.0 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13,141.22
    $13,141.22
    Each
       $13,141.22
    Supplier Number: 24406-28
  • CELLROLL Interface Cable
    Discontinued
    Supplier Number: -
  • CELLROLL Power Supply
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,365.43
    $1,365.43
    Prix promotionnel pour les nouveaux clients
    Each
       $1,365.43
    Supplier Number: 24406-31
  • CELLROLL Roller Deck
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,183.66
    $5,183.66
    Each
       $5,183.66
    Supplier Number: 13050-20
  • CELLROLL Storage Deck
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,099.71
    $1,099.71
    Prix promotionnel pour les nouveaux clients
    Each
       $1,099.71
    Supplier Number: 24406-29