DirectMark™ Laser Cassette Marker, General Data | Avantor
DirectMark™ Laser Cassette Marker, General Data
Installation/Training/Travel and Expenses Package
Catalog # CA75801-404
Fournisseur #:  IT-400/TRAVEX
DirectMark™ Laser Cassette Marker, General Data
Catalog # CA75801-404
DirectMark™  Laser Cassette Marker, General Data
DirectMark™ Laser Cassette Marker, General Data
Installation/Training/Travel and Expenses Package
Price
$29,842.20
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Reliable laser marking technology delivers enhanced throughput and accuracy.
  • Improved sample management and patient traceability maximizes lab flow efficiencies
  • Unique phase change technology permanently marks directly on cassette surface
  • Rugged and compact design features a small footprint ideal for a crowded lab
  • Three-sided printing capability provides more room for critical information
  • Full LIS/LIMS system integration
  • Maximum capacity of eight magazines holding 40 cassettes
Permanent and reliable identification of tissue samples is imperative for an efficient lab. Misidentifications can cause problems thoughout the diagnostic process, not to mention precious time wasted. The DirectMark™ Laser Cassette Marker uses an innovative laser technology to mark cassettes with high-resolution text and barcodes that can be reliably scanned and read throughout the lab's process, from grossing through archiving.
Spécifications
  • Désignation:
    Installation/Training/Travel and Expenses Package